Learn More
Medchemexpress LLC 7-(2-aminoethyl)camptothecin TFA | 99.6% | 505.44 g/mol | C24H22F3N3O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY159162A10MG
Encompass_Preferred
7-(2-aminoethyl)camptothecin TFA is the trifluoroacetic acid salt of a camptothecin derivative used as a research reagent and synthetic intermediate for preparing triple-helix-forming oligonucleotide-camptothecin (TFO-CPT) conjugates and for DNA cleavage studies.
  • High purity: 99.6% as supplied.
  • Molecular weight 505.44 g/mol; formula C24H22F3N3O6.
  • Solid form, soluble in DMSO for stock solutions.
  • Available in small research quantities such as 10 MG for synthesis and analytical work.
Catalog No. 50-002-87909