Learn More
Medchemexpress LLC 7-[3-(tert-butylamino)-2-hydroxy-propoxy]-3H-isobenzofuran-1-one hydrochloride | 55104-39-7 | 98.8% | 315.79 | C15H22ClNO4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1066671MG
Encompass_Preferred
DL 071IT is a potent, non-selective beta-adrenergic receptor blocker with intrinsic sympathomimetic and weak membrane-stabilizing activity. It reduces exercise heart rate and systolic blood pressure, and can significantly lower resting heart rate. Supplied as a hydrochloride salt for research use only.
  • Non-selective beta-adrenergic receptor blocker.
  • Exhibits intrinsic sympathomimetic activity.
  • Shows weak membrane-stabilizing activity.
  • Lowers exercise heart rate and systolic blood pressure.
  • Supplied as a hydrochloride salt, suitable for compound studies.
  • High purity (≈98.8%) for research applications.
  • Stable under recommended storage conditions.
Catalog No. 50-002-42032