Learn More
Cayman Chemical 7 8 9 10Tetrahydrobenzoapy
SDP
Supplier:  Cayman Chemical 16225500 MG
Encompass_Preferred
A benzopyrene derivative that is activated by hepatic cytosol into electrophilic sulfuric acid esters, which are capable of forming covalent DNA adducts and inducing mutations
Catalog No. 50-297-0326