Learn More
Cayman Chemical 7 8-DIHDTE 100ug
SDP
Supplier:  Cayman Chemical 20332100 UG
Encompass_Preferred
An oxylipin; oxidative metabolite of DPA; formed via CYP metabolism of DPA to the epoxide followed by hydrolysis to the diol by epoxide hydrolase
Catalog No. 50-298-3126