Learn More
Medchemexpress LLC 7,8-dihydro-2,5(1H,6H)-quinolinedione | 15450-69-8 | MFCD00766821 | 99.9% | 163.17 g/mol | C9H9NO2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW017764
Encompass_Preferred
7,8-Dihydro-2,5(1H,6H)-quinolinedione is a small-molecule quinolinedione reported as a positive inotropic agent for research applications. It is supplied as a characterized solid reagent suitable for biochemical and pharmacological studies.
  • Reported positive inotropic activity.
  • Molecular formula C9H9NO2; molecular weight 163.17 g/mol.
  • High purity (~99.9%) as supplied.
  • Solid; light brown to light yellow in appearance.
  • Offered in multiple pack sizes for laboratory use.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-003-02325