Learn More
Medchemexpress LLC 7,8-Dihydro-L-biopterin | 6779-87-9 | 96.5% | 239.23 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW008646R10MG
Encompass_Preferred
7,8-Dihydro-L-biopterin is a NOS uncoupling inducer with blood-brain barrier permeability, and it is a reduced non-conjugated pteridine. It is the main metabolite of 4-amino-tetrahydro-L-biopterin and undergoes photooxidation to form biopterin. It promotes the conversion of nitric oxide synthase to a superoxide-producing form, increasing oxidative stress levels in the renal outer medulla and inducing apoptosis.
  • Nos uncoupling inducer
  • Blood-brain barrier permeability
  • Reduced non-conjugated pteridine
  • Main metabolite of 4-amino-tetrahydro-L-biopterin
  • Sensitive to the inhibitory effect of SOD
Catalog No. 50-004-52694