Learn More
Medchemexpress LLC 7,8-Dihydro-L-biopterin (Standard) | 6779-87-9 | 99.8% | 239.23 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW00864610MG
Encompass_Preferred
7,8-Dihydro-L-biopterin is a NOS uncoupling inducer with blood-brain barrier permeability, and it is a reduced non-conjugated pteridine. It is the main metabolite of 4-amino-tetrahydro-L-biopterin, and it undergoes photooxidation to form biopterin. It promotes the conversion of nitric oxide synthase to a superoxide-producing form, thereby increasing oxidative stress levels in the renal outer medulla and inducing apoptosis. It is sensitive to the inhibitory effect of SOD.
  • Can be applied to research related to salt-sensitive hypertension.
  • Can be applied to research related to moderate to severe traumatic brain injury.
  • Can be applied to research related to neurodegenerative diseases.
Catalog No. 50-003-66832