Learn More
Cayman Chemical 7 8-DIhydronpterIn 10mg
SDP
Supplier:  Cayman Chemical 2760610 MG
Encompass_Preferred
A pteridine with antioxidant activities; scavenges DPPH radicals in a cell-free assay at 20-100 µM; inhibits copper ion-induced oxidation of LDL in a cell-free assay and decreases TBARS produced by THP-1 cells cultured with LDL at 2-10 µM; inhibits necrosis induced by oxLDL in U937, but not THP-1, cells at 200 µM
Catalog No. 50-293-7781