Learn More
Selleck Chemical LLC 7,8-Dihydroxyflavone 20mg 38183-03-8 7,8-DHF
SDP
Supplier:  Selleck Chemical LLC S831920MG
Encompass_Preferred
7,8-Dihydroxyflavone (7,8-DHF) acts as a potent and selective small-molecule agonist of the TrkB receptor (Kd ? 320 nM), the main signaling receptor of brain-derived neurotrophic factor (BDNF). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5939