Learn More
Medchemexpress LLC HY-W013372 50mg Medchemexpress, 7,8-Dihydroxyflavone CAS:38183-03-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01337250MG
Encompass_Preferred
Medchemexpress, HY-W013372 50mg 7,8-Dihydroxyflavone CAS:38183-03-8 7,8-Dihydroxyflavone is a potent and selective TrkB agonist that mimics the physiological actions of Brain-derived neurotrophic factor (BDNF). Displays therapeutic efficacy toward various neurological diseases [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4582