Learn More
Cayman Chemical 7 8EpDPA Specialty Organic Che
SDP
Supplier:  Cayman Chemical 10465500 UG
Encompass_Preferred
An epoxide derivative of DHA that is generated by the action of CYP450 epoxygenases; naturally occurring in plasma and brain and spinal cord tissues and is increased following dietary supplementation with ω-3 fatty acids; a substrate of soluble epoxide hydrolase (KM = 15 µM)
Catalog No. 50-288-9685