Learn More
Medchemexpress LLC 7-Amino-4-methylcoumarin | 26093-31-2 | 1 G
SDP
Supplier:  Medchemexpress LLC HYD00271G
Encompass_Preferred
7-Amino-4-methylcoumarin, a compound of the coumarin class, can be isolated from the endophytic fungus *Xylaria sp.* It exhibits broad-spectrum antibacterial activity and is commonly used as an important laser dye that emits in the blue region. It is capable of analyzing glycoprotein monosaccharides and N-linked oligosaccharides.
  • Utilized in tissue pathology analysis, enzyme activity measurement, and copper ion detection.
  • Excitation wavelength: 351 nm
  • Emission wavelength: 430 nm
  • Exhibits broad-spectrum antibacterial activity.
Catalog No. 50-004-10028