Learn More
Medchemexpress LLC 7-Amino-4-methylcoumarin | 26093-31-2 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYD0027500MG
Encompass_Preferred
7-Amino-4-methylcoumarin belongs to the coumarin class and can be isolated from the endophytic fungus Xylaria sp., exhibiting broad-spectrum antibacterial activity. This compound is commonly used as an important laser dye that emits in the blue region. It is capable of analyzing glycoprotein monosaccharides and N-linked oligosaccharides, and is also utilized in tissue pathology analysis, enzyme activity measurement, and copper ion detection. The excitation wavelength for 7-Amino-4-methylcoumarin is 351 nm, and its emission wavelength is 430 nm.
  • Used as an important laser dye that emits in the blue region.
  • Capable of analyzing glycoprotein monosaccharides and N-linked oligosaccharides.
  • Utilized in tissue pathology analysis.
  • Used for enzyme activity measurement.
  • Suitable for copper ion detection.
  • Exhibits broad-spectrum antibacterial activity.
Catalog No. 50-004-10025