Learn More
Medchemexpress LLC 7-Amino-4-methylcoumarin-3-acetic acid | 106562-32-7 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYD0025
Encompass_Preferred
7-Amino-4-methylcoumarin-3-acetic acid is a fluorescent protein labeling agent that emits in the blue region (440-460 nm) when activated with UV light (350 nm). It is intended for research use only.
  • Fluorescent protein labeling agent.
  • Emits in the blue region (440-460 nm).
  • Activated with UV light (350 nm).
  • Suitable for immunofluorescence and flow cytometry.
  • Used in biological conjugation and labeling chemistry.
Catalog No. 50-004-12645