Learn More
Enzo Life Sciences 7-Aminoactinomycin D (1mg). CAS: 7240-37-1
SDP
Supplier:  Enzo Life Sciences BMLAP4000001
Encompass_Preferred
Fluorescent, intercalating DNA-binding agent useful for identifying apoptotic cells by flow cytometry. Capable of identifying populations representing live, apoptotic, and late apoptotic/dead cells in a quick, simple, reproducible, cost-effective fashion. Inhibitor of DNA-primed RNA polymerase and DNA polymerase. Cytochemical probe. Exhibits growth-inhibitory activity against certain leukemias and sarcomas. has antibacterial properties. Alternative name: 7-AAD, 7AAD. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (20mg/ml), methanol or 100% ethanol. Slightly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0317