Learn More
Cayman Chemical 7-AmInoactInomycIn D 5mg
SDP
Supplier:  Cayman Chemical 113975 MG
Encompass_Preferred
A fluorescent DNA dye; commonly used for the detection or exclusion of non-viable cells in flow cytometric analysis; ex/em = 488, 546, 578/650 nm, respectively; exhibits minimal spectral overlap with commonly used probes, therfore it can be used in conjunction with probes such as FITC; has been used to evaluate apoptosis and cell-mediated cytotoxicity and to stain DNA in cells that have been fixed and permeabilized by a variety of methods
Catalog No. 50-295-1162