Learn More
Cayman Chemical 7-AmInocephalospornIc AcId 25g
SDP
Supplier:  Cayman Chemical 3582425 G
Encompass_Preferred
A cephalosporin antibiotic building block; binds to Hsp90β (Kd = 6.201 µM); inhibits SREBP activation in a reporter assay using Hl 7702 cells at 40 µM; inhibits palmitic acid- and oleic acid-induced increases in total cholesterol and triglyceride levels in HepG2 cells at 40 µM; reduces hepatic steatosis in mice fed a high-fat diet from 5-25 mg/kg; inhibits the growth of M. leprae in mice at 10 mg/kg; a potential impurity in commercial preparations of cefoperazone
Catalog No. 50-291-2438