Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Aminoheptanoic acid | 929-17-9 | 145.20 | 50 G
SDP

Supplier:  Medchemexpress LLC HYW00204250G

Encompass_Preferred

7-Aminoheptanoic acid is a PROTAC linker, which can be utilized in the synthesis of PROTAC molecules like HL-8 (HY-143275). It is for research use only and not intended for medical applications.

  • PROTAC linker
  • Used to synthesize PROTAC molecules
  • For research use only
  • Molecular weight: 145.20
  • Molecular formula: C7H15NO2
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: 3 years at room temperature; 2 years at -80°C in solvent; 1 year at -20°C in solvent

Catalog No. 50-004-47100


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.