Learn More
Medchemexpress LLC 7-azido-4-methylcoumarin | 95633-27-5 | 98.9% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11932310MG
Encompass_Preferred
7-Azido-4-methylcoumarin is a coumarin-based fluorogenic probe used for sensitive fluorescence detection of hydrogen sulfide (H2S) and as an assay substrate in enzymatic studies. The azide functional group is selectively reduced by H2S to produce a fluorescent coumarin, enabling quantitative and qualitative analysis in biochemical assays.
  • Fluorogenic probe for hydrogen sulfide (H2S) detection.
  • Azide reduction yields a strong fluorescent signal for sensitive assays.
  • Used as a substrate for cystathionine β-synthase activity measurements.
  • Supplied in small laboratory pack sizes suitable for analytical use.
  • Molecular weight 201.18 g·mol⁻¹; formula C10H7N3O2.
Catalog No. 50-002-33069