Learn More
Medchemexpress LLC 7-Bromoquinoline | 4965-36-0 | 208.06 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW004432
Encompass_Preferred
7-Bromoquinoline is a biochemical reagent that serves as an organic compound for various life science research applications. It has a high purity of 99.95%.
  • Used as a biological material
  • Used as an organic compound for life science related research
  • Molecular formula: C9H6BrN
  • Appears as a solid below 32°C and liquid above 36°C
  • Light yellow to yellow color
Catalog No. 50-004-48196