Learn More
Medchemexpress LLC 7-chloro-4-hydroxyquinoline-2-carboxylic acid | 18000-24-3 | MFCD00069227 | 99.9% | 223.61 g/mol | C10H6ClNO3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10081150MG
Encompass_Preferred
7-Chlorokynurenic acid is a research-grade small molecule used to probe NMDA receptor function and glutamatergic signaling. It acts as a potent antagonist at the glycine B coagonist site and is provided in high-purity solid and solution formats for laboratory use.
  • Selective antagonist at the NMDA receptor glycine B coagonist site.
  • High listed purity suitable for research and analytical applications.
  • Available as solid or pre-made solution for experimental flexibility.
  • Offered in small pack sizes suitable for biochemical and neuroscience assays.
  • Applicable to studies of glutamate transport, synaptic function, and nociception.
Catalog No. 50-002-11773