Learn More
Medchemexpress LLC 7-chlorokynurenic acid sodium salt | 1263094-00-3 | 99.9% | 245.59 g/mol | C10H5ClNNaO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY100811A5MG
Encompass_Preferred
7-Chlorokynurenic acid sodium salt is the sodium salt form of 7-chlorokynurenic acid, a selective antagonist at the glycine co-agonist site of NMDA receptors used in neuropharmacology research. Supplied as a high-purity solid, it is appropriate for in vitro and in vivo studies probing NMDA receptor function and glycine-site modulation.
  • High purity suitable for research applications.
  • Selective antagonist of NMDA receptor glycine site.
  • Solid sodium salt form for improved aqueous solubility.
  • Available in small, precise quantities for dose-response studies.
  • Documented analytical data and purity specification provided.
Catalog No. 50-002-11778