Learn More
Medchemexpress LLC 7-chlorokynurenic acid sodium salt | 1263094-00-3 | MFCD12195840 | >99.0% | 245.59 | C10H5ClNNaO3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY100811A50MG
Encompass_Preferred
7-Chlorokynurenic acid sodium salt is the sodium salt form of a potent and selective antagonist of the glycine co-agonist site on the NMDA receptor. Supplied for research use, the compound is intended for biochemical and pharmacological studies where selective NMDA modulation and water-soluble formulations are required.
  • Potent and selective antagonist of the NMDA glycine site.
  • Sodium salt form with improved aqueous solubility.
  • High purity suitable for research applications.
  • Soluble in water and DMSO, facilitating preparation of stock solutions.
  • Recommended short-term storage at 4°C under inert atmosphere; long-term storage in solvent at -80°C.
Catalog No. 50-002-13428