Learn More
Medchemexpress LLC 7-chlorokynurenic acid sodium salt | 1263094-00-3 | MFCD12195840 | 99.9% | 245.59 g/mol | C10H5ClNNaO3 | 1 ML

Supplier: Medchemexpress LLC HY100811A10MM 1ML
7-Chlorokynurenic acid sodium salt is a selective antagonist of the glycine B co-agonist site of the NMDA receptor. It is supplied as a ready-to-use 10 mM solution (1 mL) in DMSO for research use, with high reported purity and recommended storage guidance for both solid and solution forms.
- Selective antagonist of the glycine B co-agonist site on the NMDA receptor.
- Ready-to-use 10 mM solution in DMSO, 1 mL volume.
- High purity suitable for analytical and biological applications.
- Soluble in DMSO and water for flexible formulation.
- Molecular weight approximately 245.59 g/mol for dosing calculations.
- Storage recommendations provided for solid and solution to preserve stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.