Learn More
Medchemexpress LLC 7-cyclopentyl-2-[4-(8-isothiocyanatooctanoylamino)anilino]-N,N-dimethylpyrrolo[2,3-d]pyrimidine-6-ca | 2369981-71-3 | 98.1% | 547.71 | C29H37N7O2S | 50MG
SDP
Supplier:  Medchemexpress LLC HY12625150MG
Encompass_Preferred
CDK9-IN-7 is a selective, potent cyclin-dependent kinase 9 (CDK9) inhibitor (compound 21e) used for cell-cycle and apoptosis research. It exhibits low-nanomolar inhibitory activity and is supplied as a high-purity research reagent in powder and solution formats.
  • Selective CDK9 inhibition with IC50 ≈ 11 nM.
  • Low nanomolar potency suitable for in vitro and in vivo studies.
  • High purity (≈98.1%).
  • Available as powder and as a 10 mM solution in DMSO.
  • Multiple package sizes, including a 50 mg option.
  • Recommended storage: powder at -20°C (long term) or 4°C; in solvent at -80°C for extended storage.
Catalog No. 50-002-35958