Learn More
Medchemexpress LLC 7-deaza-2',3'-dideoxyguanosine | 111869-49-9 | 98.0% | C11H14N4O3 | 10MG

Supplier: Medchemexpress LLC HY13859210MG
7-Deaza-2',3'-dideoxyguanosine is a dideoxynucleoside research compound that inhibits HIV-1 reverse transcriptase (reported Ki ≈ 25 nM). Supplied as a solid for biochemical and antiviral assays, it is commonly used in studies of nucleotide analogs and enzyme inhibition mechanisms.
- High purity: 98.0% by HPLC.
- Solid white to off-white appearance.
- Molecular formula C11H14N4O3; MW 250.25 g·mol⁻¹.
- Available in small research pack sizes (mg scale).
- Storage: protect from light; dry storage 4°C; in solvent -80°C (6 months), -20°C (1 month).
- Suitable for biochemical assays and mechanism-of-action studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.