Learn More
Medchemexpress LLC 7-deaza-2'-deoxy-7-iodoadenosine | 166247-63-8 | MFCD07778650 | 99.7% | 376.15 g·mol⁻¹ | C11H13IN4O3 | 100 MG

Supplier: Medchemexpress LLC HYW048490100MG
7-Deaza-2'-deoxy-7-iodoadenosine is an iodinated deoxynucleoside used as a synthetic intermediate and research reagent for nucleoside chemistry. It is supplied for research use only.
- Iodinated deoxynucleoside intermediate for synthetic chemistry.
- Molecular formula C11H13IN4O3 and molecular weight 376.15 g·mol⁻¹.
- Purity reported approximately 99.6-99.7% depending on documentation.
- Typical package size: 100 MG (in-stock option).
- Supplied as a research reagent for laboratory applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.