Learn More
Medchemexpress LLC 7-Deazaguanine | 7355-55-7 | 99.7% | 150.14 | 10 G
SDP
Supplier:  Medchemexpress LLC HY3422210G
Encompass_Preferred
7-Deazaguanine is described as a highly selective, well-tolerated, and brain-penetrant DYRK1A inhibitor. It holds promise for research related to cancers and Down's syndrome.
  • Highly selective DYRK1A inhibitor.
  • Well-tolerated.
  • Brain-penetrant.
  • Promising for research in cancers and Down's syndrome.
Catalog No. 50-004-04094