Learn More
Medchemexpress LLC 7-Deazahypoxanthine | 3680-71-5 | 99.9% | C6H5N3O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY7754310MM/1ML
Encompass_Preferred
7-Deazahypoxanthine is a biochemical reagent designed for life science research. This product is supplied as a 10 mM solution in DMSO. It serves as a biological material or organic compound and is also utilized as a pharmaceutical intermediate. Its methylated forms are known for their use as regiochemical probes of xanthine oxidase.
  • Biochemical reagent for life science related research.
  • Can be used as a biological material or organic compound.
  • Used as a pharmaceutical intermediate.
  • Methylated derivatives act as regiochemical probes of xanthine oxidase.
Catalog No. 50-004-35084