Learn More
Cayman Chemical 7-DemethylsuberosIn 10mg
SDP
Supplier:  Cayman Chemical 3528210 MG
Encompass_Preferred
A prenylated coumarin with anti-inflammatory and neuroprotective activities; inhibits TPA- and A23187-induced histamine release from HMC-1 cells at 10 µM; protects against glutamate-induced cytotoxicity in primary rat cortical cells at 0.1-10 µM
Catalog No. 50-291-0001