Learn More
Medchemexpress LLC 7-diethylamino-3-thenoylcoumarin | 77820-11-2 | MFCD00051376 | 99.2% | 327.40 g·mol⁻¹ | C18H17NO3S | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW150677250MG
Encompass_Preferred
BioA-IN-1 is a small-molecule inhibitor of BioA, an enzyme in the biotin biosynthesis pathway of Mycobacterium tuberculosis. Reported as compound 15 in the literature, it has an IC50 of 0.195 μM and shows antibacterial activity with no reported cytotoxicity; it is supplied for research use.
  • Inhibits BioA enzyme involved in biotin biosynthesis.
  • Reported IC50 of 0.195 μM (compound 15).
  • Demonstrates antibacterial activity without reported cytotoxicity.
  • High reported purity (≈99.2%).
  • Chemical formula C18H17NO3S; molecular weight 327.40 g·mol⁻¹.
  • Available in small-scale packaging suitable for research (e.g., 250 mg).
Catalog No. 50-002-83148