Learn More
Cayman Chemical 7 DIethylamInocoumarIn3ca
SDP
Supplier:  Cayman Chemical 35396500 MG
Encompass_Preferred
A fluorescent probe; has been used as a FRET acceptor conjugated to neomycin B or tobramycin for use in the analysis of bacterial RNA binding; has been used as a reactive probe for the derivatization of sulfonamides for LIF detection; ex/em = 407/472 nm, respectively
Catalog No. 50-291-3364