Learn More
Apexbio Technology LLC 7-Dimethylaminocoumarin-4-acetic acid 80883-54-1 100mg
SDP
Supplier:  Apexbio Technology LLC A4719100
Encompass_Preferred
7-Dimethylaminocoumarin-4-acetic acid (CAS 80883-54-1) is a fluorescent probe used to visualize or assess enzymatic activities and signaling pathway dynamics at the cellular level Its coumarin-derived structure with a dimethylamino group enables fluorescence facilitating biological labeling imaging and diagnostic research 7-Dimethylaminocoumarin-4-acetic acid exerts its biological activity primarily through interaction with specific biological targets allowing for monitoring of cellular functions and pathway interactions relevant to disease progression studies and therapeutic development Based on these properties 7-Dimethylaminocoumarin-4-acetic acid holds research potential in cellular imaging enzymatic assays and diagnostic analyses
Catalog No. 50-000-93468