Learn More
Cayman Chemical 7-epI-10-Deacetyltaxol 10mg
SDP
Supplier:  Cayman Chemical 3429610 MG
Encompass_Preferred
A taxane diterpenoid with α-glucosidase inhibitory and anticancer activities; inhibits α-glucosidase (IC50 = 48.8 µM) and is cytotoxic to HeLa cells (IC50 = 0.085 nM); induces apoptosis and DNA fragmentation in HepG2 cells
Catalog No. 50-294-8064