Learn More
Medchemexpress LLC 7-Epi 10-desacetyl paclitaxel | 78454-17-8 | 99.8% | C45H49NO13 | 5 MG

Supplier: Medchemexpress LLC HY775725MG
7-Epi 10-desacetyl paclitaxel is a taxol derivative that demonstrates cytotoxicity against HeLa cells with an IC50 of 85 μM. This product is intended for research use only.
- Exhibits cytotoxicity against HeLa cells (IC50 of 85 μM)
- Provided as a white to yellow solid
- Suitable for research applications
- High purity (99.8%)
- Molecular weight of 811.87
- Chemical formula C45H49NO13
- Identified by CAS number 78454-17-8
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.