Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-ethyl-4-[3-(4-ethylsulfonyl-2-methoxyphenyl)-4-fluorophenyl]imidazo[4,5-c]pyridazine | 1614245-70-3 | 99.3% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY12087410MG

Encompass_Preferred

Darigabat is a subtype-selective GABAA positive allosteric modulator used for preclinical research into epilepsy, anxiety, and seizure disorders. It is a small-molecule research compound with confirmed chemical identity and supplier-stated purity.

  • CAS number 1614245-70-3.
  • Chemical formula C22H21FN4O3S.
  • Molecular weight 440.49 g/mol.
  • Purity 99.3% (supplier listing).
  • Packaging 10 MG; solid storage 4°C, protect from light; in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-30936


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.