Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Fluorobenzofurazan-4-sulfonic acid ammonium | 84806-27-9 | 98.96% | 50 MG
SDP

Supplier:  Medchemexpress LLC HYD009750MG

Encompass_Preferred

7-Fluorobenzofurazan-4-sulfonic acid ammonium is a fluorescent label that can be utilized for detecting dissolved thiol-disulfide, such as Cys and GSH. It is intended for research use only and not for patient use.

  • Fluorescent label: can be used for detecting dissolved thiol-disulfide (e.g., Cys, GSH).
  • Purity: 98.96%.
  • Appearance: solid.
  • Color: light yellow to yellow.
  • Emission (Em): 495.
  • Excitation (Ex): 478.
  • Solubility: soluble in DMSO (250 mg/mL) and H2O (≥ 50 mg/mL).
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light).

Catalog No. 50-004-10668


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.