Learn More
American Radiolabeled Chemicals Inc 7-HYCOUMARIN 3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 177550UC
Encompass
7-Hydroxycoumarin-ring-3H 50uCi in ethanol, 1 mCi/ml, S.A. 10-30 Ci/mmol, M.W. 162.14, Shipped in Dry Ice
Catalog No. 50-901-113
Add to Cart
Request a Quote