Learn More
Medchemexpress LLC 7-hydroxy-4-phenylcoumarin | 2555-30-8 | MFCD00024080 | 96.9% | 238.24 g/mol | C15H10O3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW207195500MG
Encompass_Preferred
7-Hydroxy-4-phenylcoumarin is a coumarin derivative used as a research reagent that functions as a dual inhibitor of aldehyde dehydrogenase-2 (ALDH-2) and monoamine oxidase (MAO). It exhibits reported IC50 values of 1.5 μM (ALDH-2) and 0.5 μM (MAO) and is supplied as a solid and as DMSO solutions at high purity.
  • Dual ALDH-2 and MAO inhibitor with low-micromolar potency.
  • High purity solid suitable for research applications.
  • Available in multiple pack sizes, including 500 mg solid.
  • Also supplied as a ready-to-use 10 mM solution in DMSO.
  • Molecular formula C15H10O3; molecular weight 238.24 g/mol.
Catalog No. 50-002-82488