Learn More
Medchemexpress LLC 7-Hydroxycarbostyril | 70500-72-0 | 99.1% | 161.16 | 10 G
SDP
Supplier:  Medchemexpress LLC HY3476310G
Encompass_Preferred
7-Hydroxycarbostyril (synonyms: 7-Hydroxyquinolinone; 2,7-Dihydroxyquinoline) is a quinolinone derivative. It can be used for the synthesis of Brexpiprazole, an orally active antipsychotic agent that acts as a partial agonist of human 5-HT1A and dopamine D2L receptors. This product is for research use only and not sold to patients.
  • Quinolinone derivative
  • Used for synthesis of Brexpiprazole
  • Brexpiprazole acts as a partial agonist of human 5-HT1A and dopamine D2L receptors
  • For research use only
Catalog No. 50-004-04249