Learn More
Medchemexpress LLC 7α-Hydroxycholesterol | 566-26-7 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN726410MM/1ML
Encompass_Preferred
7α-Hydroxycholesterol is a cholesterol oxide and can serve as a biomarker for oxidative stress and lipid peroxidation. It has cytotoxic and pro-inflammatory activities, and can inhibit sterol synthesis and reduce the activity of HMG-CoA reductase. 7α-Hydroxycholesterol can be used in the research of diseases such as diabetes and atherosclerosis. It is for research use only and not sold to patients.
  • Biomarker for oxidative stress and lipid peroxidation
  • Cytotoxic and pro-inflammatory activities
  • Inhibits sterol synthesis and reduces HMG-CoA reductase activity
  • Applicable in diabetes and atherosclerosis research
Catalog No. 50-004-18019