Learn More
Medchemexpress LLC 7-hydroxymethotrexate | 5939-37-7 | MFCD00674903 | 98.1% | C20H22N8O6 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13056910MG
Encompass_Preferred
7-Hydroxymethotrexate is the major oxidative metabolite of methotrexate provided as a research-grade analytical standard for metabolic and pharmacokinetic studies. Key identifiers include CAS 5939-37-7, molecular formula C20H22N8O6, molecular weight 470.44 g·mol⁻1, and reported purity 98.14%.
  • High purity for analytical and metabolic studies.
  • Available in small milligram pack sizes for research use.
  • Characterized by molecular formula C20H22N8O6 and molecular weight 470.44 g·mol⁻1.
  • Supplied with supporting product information and datasheet for traceability.
  • Suitable as a reference standard in pharmacokinetic and biochemical assays.
Catalog No. 50-002-60938