Learn More
TARGETMOL CHEMICALS INC 7-keto-deoxycholic acid 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN712025MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. 7-keto-deoxycholic acid is converted from Lactobacillus and Bifidobacterium with specific condition and from cholic acid in vitro. Purity 98.76%
Catalog No. 50-288-0257