Learn More
Medchemexpress LLC 7-keto-Deoxycholic acid | 911-40-0 | 99.7% | 406.56 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY413245MG
Encompass_Preferred
7-keto-Deoxycholic acid is a metabolite of bile acids found in *Clostridium absonum*. It is also produced from *Lactobacillus* and *Bifidobacterium* under specific conditions. In vitro, its bioconversion rate increases with the addition of an equimolar concentration of deoxycholic acid. It is induced by deoxycholic acid and subsequently metabolized. For example, *E. lentum-like c-25* can convert cholic acid to deoxycholic acid and 7-keto-Deoxycholic acid under anaerobic incubation.
  • Metabolite of bile acids.
  • Can be converted from *Lactobacillus* and *Bifidobacterium*.
  • Available in various quantities.
  • High purity.
  • Solid appearance, white to off-white color.
  • Endogenous metabolite.
  • Shipping at room temperature in continental US; storage at 4°C, protected from light.
  • Soluble in DMSO.
Catalog No. 50-004-03644