Learn More
Medchemexpress LLC 7-ketolithocholic acid | 4651-67-6 | MFCD00271393 | 390.6 g/mol | C24H38O4 | 10 MG

Supplier: Medchemexpress LLC HYW018512R10MG
7-Ketolithocholic acid is a bile acid derivative supplied as an analytical standard for research and analytical applications. It is reported to be absorbable and to suppress endogenous bile acid production and biliary cholesterol secretion, making it useful in metabolic and analytical studies.
- Used as an analytical standard for bile acid research.
- Provides a defined reference for chromatographic and mass spectrometry assays.
- Molecular formula C24H38O4 and molecular weight 390.6 g/mol support mass calculation and identifications.
- CAS number 4651-67-6 enables unambiguous compound identification.
- Intended for research use only; not for human therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.