Learn More
AdipoGen 7-Methoxycoumarin-acetic acid
SDP
Supplier:  AdipoGen CDXM0028G001
Encompass_Preferred
Chemical. CAS 62935-72-2. Formula C12H10O5. MW 234.2. Synthetic Florescent probe that is commonly used as a building block to synthesize coumarin-type fluorescent probes. Spectral data lambdaex 320 nm, lambdaem 380 nm in methanol. It is used as a fluorescent label for peptides. It is widely used for developing FRET peptide substrates for analyzing protease activities and for HPLC derivatization by fluorescence detection.
Catalog No. 50-256-0558