Learn More
Cayman Chemical 7-MethoxyIsoflavon 5g
SDP
Supplier:  Cayman Chemical 334195 G
Encompass_Preferred
A synthetic isoflavone; inhibits concanavalin A-induced mitosis in PBMCs (IC50 = 1.36 UG/ml); inhibits the growth of HCT116 colon cancer cells at 10, 20, and 40 µM
Catalog No. 50-294-6685