Learn More
Medchemexpress LLC 7-Methoxyrosmanol | 113085-62-4 | 99.3% | 360.44 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1118961MG
Encompass_Preferred
7-Methoxyrosmanol is a phenolic diterpene isolated from rosemary that suppresses the cAMP responsiveness of PEPCK and G6Pase promoters. This product is for research use only.
  • Phenolic diterpene
  • Suppresses cAMP responsiveness of PEPCK and G6Pase promoters
  • Available in various sizes
  • Available in solid and solution forms
Catalog No. 50-004-38208