Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene | 84030-20-6 | 99.19% | C8H15N3 | 10 G
SDP

Supplier:  Medchemexpress LLC HY5915710G

Encompass_Preferred

7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene is a drug intermediate, primarily used for the synthesis of various active compounds. It is supplied as a colorless to light yellow liquid. This product is intended for research use only.

  • Drug intermediate for synthesis of various active compounds
  • Colorless to light yellow liquid appearance
  • Molecular weight of 153.23
  • Chemical formula C8H15N3

Catalog No. 50-004-34748


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.