Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Methyl-1-naphthyl acetic acid | 6836-22-2 | 216.24 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW003836

Encompass_Preferred

7-Methyl-1-naphthyl acetic acid is a potent inhibitor of auxin action in plants, specifically inhibiting AUX1, PIN, and ABCB protein-mediated auxin transport expressed in yeast. It is utilized as a laboratory chemical and in the manufacture of substances for auxin-dependent development.

  • Purity of 99.99%
  • Molecular weight of 216.24
  • Chemical formula: C13H12O3
  • Appears as a white to off-white solid
  • Melting point: 153-154°C
  • Boiling point: 408.8°C at 760 mmHg
  • Stable under recommended storage conditions
  • Functions as a potent inhibitor of auxin action
  • Supports auxin-dependent development studies
  • Recommended powder storage: -20°C for 3 years or 4°C for 2 years
  • Recommended solvent storage: -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-49540


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.